• USA Home
  • Search Results

  • camphordl152237622211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Camphor (dl)

1 Products
Synonym:   (±)-Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one
Image of Camphor (dl)
CAS Number:
Empirical Formula (Hill Notation):
 C10H16O
Molecular Weight:
 152.23
 Beilstein/REAXYS Number:
 1907611
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More