• USA Home
  • Search Results

  • camphorwhiteoil12345800851311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Camphor white oil

1 Products
Synonym:   Camphora oil, Cinnamomum Camphora Sieb
CAS Number:
 Council of Europe no.:
 0130
 FEMA Number:
 2231
 MDL number:
 NACRES:
 NA.21
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
Show More