• USA Home
  • Search Results

  • cantharidin196205625711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cantharidin

2 Products
Synonym:   3a,7a-Dimethylhexahydro-4,7-epoxyisobenzofuran-1,3-dione, 7a-Dimethylhexahydro-3a,4,7-epoxyisobenzofuran, Cantharidine, Hexahydro-3a,7a-dimethyl-4,7-epoxyisobenzofuran-1,3-dione
Image of Cantharidin
CAS Number:
Empirical Formula (Hill Notation):
 C10H12O4
Molecular Weight:
 196.20
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More