• USA Home
  • Search Results

  • caprylicacid1442112407211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Caprylic acid

2 Products
Synonym:   Acid C8, Caprylic acid, Octanoic acid
Image of Caprylic acid
CAS Number:
Linear Formula:
 CH3(CH2)6COOH
Molecular Weight:
 144.21
 Beilstein/REAXYS Number:
 1747180
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More