• USA Home
  • Search Results

  • caprylicacidoctanoicacid1442112407211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Caprylic acid (Octanoic acid)

2 Products
Synonym:   Caprylic acid (Octanoic acid)
Image of Caprylic acid (Octanoic acid)
CAS Number:
Linear Formula:
 CH3(CH2)6COOH
Molecular Weight:
 144.21
 EC Index Number:
 204-677-5
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Agency/Method
  Special Grade
  Application(s)
Show More