• USA Home
  • Search Results

  • caps22132113540611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

CAPS

4 Products
Synonym:   3-(Cyclohexylamino)-1-propanesulfonic acid
Image of CAPS
CAS Number:
Linear Formula:
 C6H11NH(CH2)3SO3H
Molecular Weight:
 221.32
 Beilstein/REAXYS Number:
 2835588
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  pK-Values
  UV-Absorption Values
  Absorption
  Product Line
  Physical Form
  pH Value
  Solubility
Show More