• USA Home
  • Search Results

  • capsaicin3054140486411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Capsaicin

10 Products
Synonym:   8-Methyl-N-vanillyl-trans-6-nonenamide
Image of Capsaicin
CAS Number:
Linear Formula:
 (CH3)2CHCH=CH(CH2)4CONHCH2C6H3-4-(OH)-3-(OCH3)
Molecular Weight:
 305.41
 Beilstein/REAXYS Number:
 2816484
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Chemical Composition
  Application(s)
  Manufacturer designation
  Biological Source
  Anal. Standard Form
  Physical Form
  Solubility
  Assay
Show More