• USA Home
  • Search Results

  • captopril217296257186211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Captopril

6 Products
Synonym:   N-[(S)-3-Mercapto-2-methylpropionyl]-L-proline
Image of Captopril
CAS Number:
Empirical Formula (Hill Notation):
 C9H15NO3S
Molecular Weight:
 217.29
 Beilstein/REAXYS Number:
 477887
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Biological Source
  Assay
  Anal. Standard Form
  Manufacturer designation
  Physical Form
Show More