• USA Home
  • Search Results

  • carbaryl201226325211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbaryl

1 Products
Synonym:   1-Naphthyl-N-methylcarbamate
Image of Carbaryl
CAS Number:
Linear Formula:
 C10H7OCONHCH3
Molecular Weight:
 201.22
 Beilstein/REAXYS Number:
 1875862
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More