• USA Home
  • Search Results

  • carbazole167218674811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbazole

4 Products
Synonym:   Diphenylenimine, 9-Azafluorene, Diphenylenimine, 9-Azafluorene
Image of Carbazole
CAS Number:
Empirical Formula (Hill Notation):
 C12H9N
Molecular Weight:
 167.21
 EC Index Number
 201-696-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
  Titertest
  Assay
Show More