• USA Home
  • Search Results

  • carbenicillindisodiumsalt42236480094611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbenicillin disodium salt

2 Products
Synonym:   α-Carboxybenzylpenicillin disodium salt
Image of Carbenicillin disodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C17H16N2Na2O6S
Molecular Weight:
 422.36
 Beilstein Registry Number:
 5722128
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Mode of Action
  Physical Form
  Solubility
Show More