• USA Home
  • Search Results

  • carbon1201744044011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon

152 Products
Synonym:   Carbon
CAS Number:
Empirical Formula (Hill Notation):
 C
Molecular Weight:
 12.01
 EC Number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Weight
  Particle Dimension
  Assay
  Physical Form
  Manufacturer designation
Show More

Product #
Description
484164 Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis,
633100 Carbon, nanopowder, <100 nm particle size (TEM),
GF23879009 Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%,
GF34902109 Carbon, foil, 10x10mm, thickness 2.0mm, hOpg,
GF52352355 Carbon, powder, max. particle size 75 micron, weight 100 g, high purity 99.997%,
GF24640950 Carbon, rod, 100mm, diameter 10.0mm, graphite, 100%,
GF01171763 Carbon, foil, 50x50mm, thickness 0.35mm, flexible graphite, 99.8%,
GF97921086 Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%,
GF14123685 Carbon, powder, max. particle size 75 micron, weight 20 g, high purity 99.997%,
GF76996984 Carbon, rod, 150mm, diameter 3.0mm, graphite, 100%,
GF22451408 Carbon, rod, 150mm, diameter 6.35mm, graphite, 100%,
GF93506171 Carbon, rod, 100mm, diameter 2.0mm, graphite, 100%,
GF87162857 Carbon, foil, 50x50mm, thickness 0.5mm, flexible graphite, 99.8%,
GF56647211 Carbon, rod, 100mm, diameter 13.0mm, graphite, 100%,
GF27179286 Carbon, foil, 150x150mm, thickness 1.0mm, flexible graphite, 99.8%,
GF60808844 Carbon, lump, 1.5 mm max. lump size, weight 100 g, purity 99.5%,
GF65371271 Carbon, lump, 1.5 mm max. lump size, weight 50 g, purity 99.5%,
GF70666825 Carbon, rod, 3.0 mm diameter, length 100 mm, purity 99.999%,
GF72504716 Carbon, lump, 1.5 mm max. lump size, weight 100 g, purity 99.8%,
GF74194623 Carbon, foil, thickness 10.0 mm, size 100 × 100 mm, purity 99.5%,
GF77042561 Carbon, rod, 3.0 mm diameter, purity 99.999%, length 150 mm,
GF81101652 Carbon, foil, thickness 10.0 mm, size 150 × 150 mm, purity 99.5%,
GF88411811 Carbon, foil, thickness 10.0 mm, size 50 × 50 mm, purity 99.5%,
GF99294272 Carbon, lump, 1.5 mm max. lump size, weight 200 g, purity 99.5%,
GF01511626 Carbon, powder, max. particle size 75 micron, weight 10 g, high purity 99.997%,
GF07302585 Carbon, foil, thickness 10.0 mm, size 48 × 150 mm, purity 99.5%,
GF09856648 Carbon, lump, 1.5 mm max. lump size, weight 500 g, purity 99.5%,
GF13592516 Carbon, foil, thickness 1.0 mm, 25 mm diameter, purity 99.99%,
GF23930894 Carbon, tube, outside diameter 3.18 mm, length 100 mm, wall thickness 0.8 mm,
GF26180030 Carbon, lump, 1.5 mm max. lump size, weight 50 g, purity 99.8%,
GF31925213 Carbon, tube, outside diameter 3.18 mm, length 150 mm, wall thickness 0.8 mm,
GF38641585 Carbon, rod, 3.0 mm diameter, length 300 mm, purity 99.999%,
GF41417669 Carbon, tube, outside diameter 3.18 mm, length 50 mm, wall thickness 0.8 mm,
GF44538295 Carbon, powder, max. particle size 75 micron, weight 50 g, high purity 99.997%,
GF53241599 Carbon, lump, 1.5 mm max. lump size, weight 1000 g, purity 99.5%,
GF55138529 Carbon, tube, outside diameter 6.35 mm, length 50 mm, wall thickness 1.6 mm,
GF13300322 Carbon, foil, 100x100mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%,
GF91212511 Carbon, foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%,
GF31110859 Carbon, foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%,
GF82433538 Carbon, foil, 100x100mm, thickness 0.2mm, flexible graphite, 99.8%,
GF46470944 Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%,
GF37715182 Carbon, foil, 100x100mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%,
GF86742120 Carbon, foil, 100x100mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%,
GF23417434 Carbon, foil, 100x100mm, thickness 1.0mm, flexible graphite, 99.8%,
GF61313600 Carbon, foil, 100x100mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%,
GF44375609 Carbon, foil, 100x100mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%,
GF11412470 Carbon, foil, 100x100mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%,
GF15163297 Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%,
GF95868365 Carbon, foil, 150x150mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%,
GF36978104 Carbon, foil, 150x150mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%,
GF04191971 Carbon, foil, 150x150mm, thickness 0.2mm, flexible graphite, 99.8%,
GF08868314 Carbon, foil, 150x150mm, thickness 0.35mm, flexible graphite, 99.8%,
GF78361285 Carbon, foil, 150x150mm, thickness 0.5mm, flexible graphite, 99.8%,
GF31912039 Carbon, foil, 150x150mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%,
GF82985844 Carbon, foil, 150x150mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%,
GF26559513 Carbon, foil, 150x150mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%,
GF09288906 Carbon, foil, 15x15mm, thickness 2.0mm, hOpg,
GF29579787 Carbon, foil, 200x200mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%,
GF08538970 Carbon, foil, 250x250mm, thickness 0.35mm, flexible graphite, 99.8%,
GF13406633 Carbon, foil, 250x250mm, thickness 0.5mm, flexible graphite, 99.8%,
GF97410708 Carbon, foil, 250x250mm, thickness 1.0mm, flexible graphite, 99.8%,
GF60685656 Carbon, foil, 25x25mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%,
GF57718503 Carbon, foil, 25x25mm, thickness 0.15mm, rigid graphite, fine grain size, 99.95%,
GF00462912 Carbon, foil, 25x25mm, thickness 0.1mm, pyrolytic graphite, 99.99%,
GF62540232 Carbon, foil, 25x25mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%,
GF27136100 Carbon, foil, 25x25mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%,
GF39666077 Carbon, foil, 25x25mm, thickness 0.2mm, flexible graphite, 99.8%,
GF41284953 Carbon, foil, 25x25mm, thickness 0.5mm, pyrolytic graphite, 99.99%,
GF01564017 Carbon, foil, 25x25mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%,
GF37688754 Carbon, foil, 25x25mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%,
GF14397521 Carbon, foil, 25x25mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%,
GF64201249 Carbon, foil, 25x25mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%,
GF23996290 Carbon, foil, 300x300mm, thickness 0.2mm, flexible graphite, 99.8%,
GF99536338 Carbon, foil, 500x500mm, thickness 0.2mm, flexible graphite, 99.8%,
GF37642822 Carbon, foil, 500x500mm, thickness 0.35mm, flexible graphite, 99.8%,
GF29446814 Carbon, foil, 500x500mm, thickness 0.5mm, flexible graphite, 99.8%,
GF76337871 Carbon, foil, 500x500mm, thickness 1.0mm, flexible graphite, 99.8%,
GF85706400 Carbon, foil, 50x50mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%,
GF04119252 Carbon, foil, 50x50mm, thickness 0.1mm, pyrolytic graphite, 99.99%,
GF17529701 Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%,
GF86694069 Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%,
GF58259469 Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%,
GF46425078 Carbon, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%,
GF95501729 Carbon, foil, 50x50mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%,
GF11093936 Carbon, foil, 50x50mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%,
GF13787332 Carbon, foil, 50x50mm, thickness 1.0mm, flexible graphite, 99.8%,
GF99690508 Carbon, foil, 50x50mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%,
GF25155008 Carbon, foil, 50x50mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%,
GF42491291 Carbon, foil, 50x50mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%,
GF78424735 Carbon, foil, 5x10mm, thickness 2.0mm, hOpg,
GF71469939 Carbon, foil, 5x5mm, thickness 2.0mm, hOpg,
GF64932947 Carbon, foil, 10mm disks, thickness 0.2mm, flexible graphite, 99.8%,
GF83177949 Carbon, foil, 12mm disks, thickness 0.5mm, rigid graphite, fine grain size, 99.95%,
GF91624103 Carbon, foil, 12mm disks, thickness 1.0mm, rigid graphite, fine grain size, 99.95%,
GF16618327 Carbon, foil, 15mm disks, thickness 0.2mm, flexible graphite, 99.8%,
GF77290162 Carbon, foil, 25mm disks, thickness 0.2mm, flexible graphite, 99.8%,
GF42041944 Carbon, foil, 4mm disks, thickness 0.2mm, flexible graphite, 99.8%,
GF61271018 Carbon, foil, 50mm disks, thickness 0.2mm, flexible graphite, 99.8%,
GF55242558 Carbon, foil, 6mm disks, thickness 0.2mm, flexible graphite, 99.8%,
GF05319468 Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%,
GF05561849 Carbon, foil, not light tested, 100x100mm, thickness 0.075mm, flexible graphite, 99.8%,
GF71541031 Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%,
GF42290939 Carbon, foil, not light tested, 150x150mm, thickness 0.125mm, flexible graphite, 99.8%,
GF02861782 Carbon, foil, not light tested, 300x300mm, thickness 0.075mm, flexible graphite, 99.8%,
GF77841906 Carbon, foil, not light tested, 300x300mm, thickness 0.125mm, flexible graphite, 99.8%,
GF31780571 Carbon, microleaf, 24x70mm, thinness 5.0μm, specific density 1000μg/cm2, 99.997%,
GF86710620 Carbon, microleaf, 25x70mm, thinness 0.0025μm, specific density 0.5μg/cm2, temporary glass support, 99.997%,
GF38827682 Carbon, microleaf, 25x70mm, thinness 0.025μm, specific density 5μg/cm2, temporary glass support, 99.997%,
GF53461100 Carbon, microleaf, 25x70mm, thinness 0.050μm, specific density 10μg/cm2, temporary glass support, 99.997%,
GF82638029 Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%,
GF29753411 Carbon, microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, 99.997%,
GF04968096 Carbon, microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, annealed, 99.997%,
GF19177397 Carbon, microleaf, 25x70mm, thinness 1.25μm, specific density 250μg/cm2, 99.997%,
GF97174840 Carbon, microleaf, 25x70mm, thinness 2.5μm, specific density 500μg/cm2, 99.997%,
GF24548463 Carbon, microleaf, 50x70mm, thinness 1.25μm, specific density 250μg/cm2, 99.997%,
GF46855930 Carbon, microleaf, 50x70mm, thinness 2.5μm, specific density 500μg/cm2, 99.997%,
GF21218802 Carbon, rod, 100mm, diameter 25mm, graphite, 100%,
GF81064076 Carbon, rod, 100mm, diameter 25mm, graphite, 99.95%,
GF49415979 Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%,
GF77888121 Carbon, rod, 100mm, diameter 5.0mm, graphite, 100%,
GF77648798 Carbon, rod, 100mm, diameter 50mm, graphite, 99.95%,
GF74245507 Carbon, rod, 100mm, diameter 6.35mm, graphite, 100%,
GF80118828 Carbon, rod, 150mm, diameter 10.0mm, graphite, 100%,
GF80901312 Carbon, rod, 150mm, diameter 13.0mm, graphite, 100%,
GF30423293 Carbon, rod, 150mm, diameter 2.0mm, graphite, 100%,
GF52351242 Carbon, rod, 150mm, diameter 25mm, graphite, 100%,
GF33008947 Carbon, rod, 150mm, diameter 25mm, graphite, 99.95%,
GF36015230 Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%,
GF06099666 Carbon, rod, 150mm, diameter 50mm, graphite, 99.95%,
GF40705512 Carbon, rod, 25mm, diameter 50mm, graphite, 99.95%,
GF46430399 Carbon, rod, 2mm, diameter 0.5mm, graphite, 99.95%,
GF14853179 Carbon, rod, 300mm, diameter 10.0mm, graphite, 100%,
GF12664380 Carbon, rod, 300mm, diameter 13.0mm, graphite, 100%,
GF70707438 Carbon, rod, 300mm, diameter 2.0mm, graphite, 100%,
GF26117636 Carbon, rod, 300mm, diameter 25mm, graphite, 99.95%,
GF24623108 Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%,
GF01904015 Carbon, rod, 300mm, diameter 5.0mm, graphite, 100%,
GF73005825 Carbon, rod, 300mm, diameter 6.35mm, graphite, 100%,
GF98973613 Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%,
GF12199608 Carbon, rod, 50mm, diameter 1.0mm, graphite, 99.95%,
GF47838752 Carbon, rod, 50mm, diameter 1.5mm, graphite, 100%,
GF54569751 Carbon, rod, 50mm, diameter 25mm, graphite, 100%,
GF83048519 Carbon, rod, 50mm, diameter 25mm, graphite, 99.95%,
GF95821195 Carbon, rod, 50mm, diameter 50mm, graphite, 99.95%,
GF67478361 Carbon, tube, graphite, 100mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%,
GF07395521 Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%,
GF92315224 Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%,
GF34795323 Carbon, tube, graphite, 150mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%,
GF24441106 Carbon, tube, graphite, 150mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%,
GF39402716 Carbon, tube, graphite, 150mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%,
GF25339822 Carbon, tube, graphite, 50mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%,
GF81659166 Carbon, tube, graphite, 50mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%,