• USA Home
  • Search Results

  • carbon13cdioxide4500111172411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon-13C dioxide

6 Products
Synonym:   13C Labeled carbon dioxide, 13CO2
Image of Carbon-<SUP>13</SUP>C dioxide
CAS Number:
Empirical Formula (Hill Notation):
 13CO2
Molecular Weight:
 45.00
 Beilstein/REAXYS Number:
 3087932
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Molecular weight
  Assay
Show More