• USA Home
  • Search Results

  • carbondisulfide76147515011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon disulfide

12 Products
Image of Carbon disulfide
CAS Number:
Empirical Formula (Hill Notation):
 CS2
Molecular Weight:
 76.14
 Beilstein/REAXYS Number:
 1098293
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Condition
  Quality clarifier
  Purification method
  Product Line
  Potency
  Explosion limit
  Autoignition temp
  Restriction
  Application(s)
  Ice Shipping
  Refractive Index
  Color
  Transition temperature
  Melting Point
  Purity Grade
  Solubility
  Density
  Special Grade
  UV-Absorption Values
  Physical Form
  Vapor pressure
Show More

Product #
Description
335266 Carbon disulfide, anhydrous, ≥99%,
180173 Carbon disulfide, ACS reagent, ≥99.9%,
270660 Carbon disulfide, for HPLC, ≥99.9%,
676918 Carbon disulfide, ACS reagent, ≥99.9%,
342270 Carbon disulfide, ReagentPlus®, low benzene, ≥99.9%,
1.02214 Carbon disulfide, for analysis EMSURE® ACS,Reag. Ph Eur,
31627 Carbon disulfide, for IR spectroscopy, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (GC),
424641 Carbon disulfide, ReagentPlus®, purified by redistillation, ≥99.9%,
1.02211 Carbon disulfide, EMPLURA®,
CX0396 Carbon Disulfide, Meets Reagent Specifications for testing USP/NF monographs OmniSolv®,
CX0397 Carbon Disulfide, low benzene OmniSolv®,
CX0395 Carbon Disulfide, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,