• USA Home
  • Search Results

  • carbondisulfidesolution76147515011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon disulfide solution

4 Products
Image of Carbon disulfide solution
CAS Number:
Empirical Formula (Hill Notation):
 CS2
Molecular Weight:
 76.14
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Refractive Index
  Agency/Method
  Anal. Standard Form
  Special Grade
  Purity Grade
  Package quantity
  Physical Form
  Density
  Application(s)
Show More