• USA Home
  • Search Results

  • carbonmesoporous1201133386411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon, mesoporous

5 Products
Synonym:   Graphitized carbon black
Image of Carbon, mesoporous
CAS Number:
Empirical Formula (Hill Notation):
 C
Molecular Weight:
 12.01
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Pore size
  Bulk Density
  Density
  Assay
  Greener Alternative Principles
  Particle Dimension
  Physical Form
  Surface Dimension
Show More