• USA Home
  • Search Results

  • carbonmonoxide280163008011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon monoxide

1 Products
Image of Carbon monoxide
CAS Number:
Empirical Formula (Hill Notation):
 CO
Molecular Weight:
 28.01
 Beilstein/REAXYS Number:
 3587264
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
295116 Carbon monoxide, ≥99.0%,