• USA Home
  • Search Results

  • carbonnanotubedoublewalled1234530806856611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon nanotube, double-walled

3 Products
Synonym:   DWNT, Double wall carbon nanotubes
Image of Carbon nanotube, double-walled
CAS Number:
 MDL number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Surface Dimension
  Physical Form
Show More