• USA Home
  • Search Results

  • carbonnanotubesheet1234530806856611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon nanotube sheet

2 Products
Synonym:   MWCNT, MWNT, Multiwall carbon nanotube
Image of Carbon nanotube sheet
CAS Number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Particle Dimension
  Miscellaneous Specifications
  Composite Dimension
Show More