• USA Home
  • Search Results

  • carbonnanotubesinglewalled1234530806856611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carbon nanotube, single-walled

14 Products
Synonym:   CNT, SWCNT, SWNT, Single wall carbon nanotubes, Tuball
CAS Number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Chemical Composition
  Greener Alternative Principles
  Bulk Density
  Surface Dimension
  Manufacturer designation
  ID clarifier
  Composite Dimension
  Physical Form
  Particle Dimension
Show More

Product #
Description
704121 Carbon nanotube, single-walled, (7,6) chirality, ≥90% carbon basis (≥99% as carbon nanotubes), 0.83 nm average diameter, Green Alternative
704148 Carbon nanotube, single-walled, (6,5) chirality, ≥90% carbon basis (≥99% as carbon nanotubes), 0.78 nm average diameter, Green Alternative
704113 Carbon nanotube, single-walled, ≥89% carbon basis (≥99% as carbon nanotubes), 0.7-1.3 nm diameter, Green Alternative
724777 Carbon nanotube, single-walled, ≥90% carbon basis (≥80% as carbon nanotubes), 1.3 nm diameter, Green Alternative
698695 Carbon nanotube, single-walled, 40-60 wt. % carbon basis, D × L 2-10 nm × 1-5 μm , bundle dimensions, Green Alternative
805033 Carbon nanotube, single-walled, carbon ≥85 %, >70% (carbon as SWNT), diam. 1.3-2.3 nm,
773735 Carbon nanotube, single-walled, (6,5) chirality, ≥95% carbon basis (≥99% as carbon nanotubes), 0.78 nm average diameter,
750522 Carbon nanotube, single-walled, 98% (Semiconducting),
750514 Carbon nanotube, single-walled, <1% Metal Catalyst,
750492 Carbon nanotube, single-walled, <3.5% Metal Catalyst,
750530 Carbon nanotube, single-walled, 98% (Metallic),
755710 Carbon nanotube, single-walled, >70% (TGA),
775533 Carbon nanotube, single-walled, ≥95% carbon basis (≥99% as carbon nanotubes), 0.84 nm average diameter,
900711 Carbon nanotube, single-walled, ≥98% carbon basis,