• USA Home
  • Search Results

  • carrageenan12345900007111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carrageenan

1 Products
Synonym:   Vegetable gelatin
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.25
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Physical Form
Show More