• USA Home
  • Search Results

  • carvacrol1502249975211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Carvacrol

6 Products
Synonym:   5-Isopropyl-2-methylphenol
Image of Carvacrol
CAS Number:
Linear Formula:
 (CH3)2CHC6H3(CH3)OH
Molecular Weight:
 150.22
 Beilstein/REAXYS Number:
 1860514
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Refractive Index
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Application(s)
  Manufacturer designation
  Agency/Method
  Special Grade
  Assay
Show More