• USA Home
  • Search Results

  • caryophylleneoxide22035113930611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(−)-Caryophyllene oxide

5 Products
Synonym:   β-Caryophyllene epoxide, (−)-Epoxycaryophyllene, (−)-Epoxydihydrocaryophyllene, (1R,4R,6R,10S)-9-Methylene-4,12,12-trimethyl-5-oxatricyclo[8.2.0.04,6]dodecane, trans-Caryophyllene oxide
Image of (−)-Caryophyllene oxide
CAS Number:
Empirical Formula (Hill Notation):
 C15H24O
Molecular Weight:
 220.35
 Beilstein/REAXYS Number:
 148213
 EC Number:
 FEMA Number:
 4085
 Flavis number:
 16.043
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More