• USA Home
  • Search Results

  • caseinfrombovinemilk12345900071911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Casein from bovine milk

6 Products
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.61
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
  Solubility
  Product Line
  Physical Form
  Biological Source
Show More