• USA Home
  • Search Results

  • casticin3743447991411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Casticin

5 Products
Synonym:   3′,5-Dihydroxy-3,4′,6,7-tetramethoxyflavone, 3,4′,6,7-Tetra-O-methylquercetagetin, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxy-4H-1-benzopyran-4-one, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxychromen-4-one, Quercetagetin 3,4′,6,7-tetramethyl ether, Vitexicarpin
Image of Casticin
CAS Number:
Empirical Formula (Hill Notation):
 C19H18O8
Molecular Weight:
 374.34
 Beilstein/REAXYS Number:
 1300392
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Physical Form
  Application(s)
  Manufacturer designation
  Assay
Show More