• USA Home
  • Search Results

  • cathepsindfrombovinespleen12345902526711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cathepsin D from bovine spleen

1 Products
CAS Number:
 Enzyme Commission (EC) Number:
 3.4.23.5  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More