• USA Home
  • Search Results

  • cefoperazonedihydrate6817011382644111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cefoperazone dihydrate

3 Products
Image of Cefoperazone dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C25H27N9O8S2 · 2H2O
Molecular Weight:
 681.70
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
Show More