• USA Home
  • Search Results

  • ceftazidimeforpeakidentification636657843906211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ceftazidime for peak identification

1 Products
Synonym:   Ceftazidime pentahydrate
CAS Number:
Empirical Formula (Hill Notation):
 C22H22N6O7S2 · 5H2O
Molecular Weight:
 636.65
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More