• USA Home
  • Search Results

  • cefuroximeaxetil510476454407611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cefuroxime Axetil

5 Products
Synonym:   [6R-[6α,7β(Z)]]-3-[[(Aminocarbonyl)oxy]methyl]-7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid 1-(acetyloxy)ethyl ester
Image of Cefuroxime Axetil
CAS Number:
Empirical Formula (Hill Notation):
 C20H22N4O10S
Molecular Weight:
 510.47
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Activity Spectrum
  Application(s)
  Assay
  Physical Form
  Manufacturer designation
Show More