• USA Home
  • Search Results

  • celastrol450613415783011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Celastrol

2 Products
Synonym:   10-Hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydro-picene-2-carboxylic acid, Tripterin
Image of Celastrol
CAS Number:
Empirical Formula (Hill Notation):
 C29H38O4
Molecular Weight:
 450.61
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More