• USA Home
  • Search Results

  • ceriumdopedironoxide1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cerium doped iron oxide

5 Products
Synonym:   Core-shell Ce3/4+ doped iron oxide nanoparticles, Core-shell type cerium doped iron oxide nanoparticles, Core-shell type cerium doped maghemite nanoparticles
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Particle Dimension
Show More