• USA Home
  • Search Results

  • ceriumiiioxalatehydrate54429anhydrousbasis1575047711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cerium(III) oxalate hydrate

1 Products
Synonym:   Cerium oxalate nonahydrate
Image of Cerium(III) oxalate hydrate
CAS Number:
Linear Formula:
 Ce2(C2O4)3 · xH2O
Molecular Weight:
 544.29 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More