• USA Home
  • Search Results

  • cerulenin223271739789611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cerulenin

1 Products
Synonym:   (2R,3S,E,E)-2,3-Epoxy-4-oxo-7,10-dodecadienamide
Image of Cerulenin
CAS Number:
Empirical Formula (Hill Notation):
 C12H17NO3
Molecular Weight:
 223.27
 Beilstein/REAXYS Number:
 4140423
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More