• USA Home
  • Search Results

  • cesiumcarbonate3258253417811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cesium carbonate

6 Products
Synonym:   Carbonic acid dicesium
Image of Cesium carbonate
CAS Number:
Linear Formula:
 Cs2CO3
Molecular Weight:
 325.82
 Beilstein/REAXYS Number:
 4546405
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Purity Grade
  Special Grade
Show More