• USA Home
  • Search Results

  • cesiumfluoride151901340013011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cesium fluoride

5 Products
Synonym:   NSC 84270
Image of Cesium fluoride
CAS Number:
Empirical Formula (Hill Notation):
 CsF
Molecular Weight:
 151.90
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Values
  Solubility
Show More