• USA Home
  • Search Results

  • cesiumtriiodide513621229772211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cesium triiodide

1 Products
Image of Cesium triiodide
CAS Number:
Linear Formula:
 CsI3
Molecular Weight:
 513.62
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More