• USA Home
  • Search Results

  • cetylalcohol242443665382411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cetyl alcohol

7 Products
Synonym:   1-Hexadecanol, Palmityl alcohol
Image of Cetyl alcohol
CAS Number:
Linear Formula:
 CH3(CH2)15OH
Molecular Weight:
 242.44
 Beilstein/REAXYS Number:
 1748475
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Vapor pressure
  Transition temperature
  Solubility
  Manufacturer designation
  Density
  Bulk Density
  Autoignition temp
  Agency/Method
  Anal. Standard Form
  Physical Form
  Application(s)
  Potency
  Melting Point
  Assay
Show More