• USA Home
  • Search Results

  • cetyltrimethylammoniumhydrogensulfate381616821407311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cetyltrimethylammonium hydrogensulfate

1 Products
Image of Cetyltrimethylammonium hydrogensulfate
CAS Number:
Linear Formula:
 CH3(CH2)15N(HSO4)(CH3)3
Molecular Weight:
 381.61
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Transmittance
  Assay
Show More