• USA Home
  • Search Results

  • chapshydrate61488anhydrousbasis33171745411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

CHAPS hydrate

3 Products
Synonym:   3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, CHAPS
Image of CHAPS hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C32H58N2O7S · xH2O
Molecular Weight:
 614.88 (anhydrous basis)
 NACRES:
 NB.22
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More