• USA Home
  • Search Results

  • ches2072910347911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

CHES

4 Products
Synonym:   2-(Cyclohexylamino)ethanesulfonic acid
Image of CHES
CAS Number:
Empirical Formula (Hill Notation):
 C8H17NO3S
Molecular Weight:
 207.29
 Beilstein Registry Number:
 2967601
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Special Grade
  pH Value
  UV-Absorption Values
  Solubility
  Absorption
  Product Line
Show More