• USA Home
  • Search Results

  • chitinasefromstreptomycesgriseus12345900106311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chitinase from Streptomyces griseus

1 Products
Synonym:   Chitodextrinase, Poly(1,4-β-[2-acetamido-2-deoxy-D-glucoside]) glycanohydrolase
Image of Chitinase from <I>Streptomyces griseus</I>
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.2.1.14  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More