• USA Home
  • Search Results

  • chitinfromshrimpshells12345139861411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chitin from shrimp shells

3 Products
Synonym:   Poly-(1→4)-β-N-acetyl-D-glucosamine
Image of Chitin from shrimp shells
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Suitability
  Special Grade
  Physical Form
Show More