• USA Home
  • Search Results

  • chitosan12345901276411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chitosan

11 Products
Synonym:   Deacetylated chitin, Poly(D-glucosamine)
Image of Chitosan
CAS Number:
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Physical Form
  Special Grade
  Chemical Composition
  Viscosity
  Biological Source
  Solubility
Show More