• USA Home
  • Search Results

  • chloralhydrate1654030217011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloral hydrate

2 Products
Synonym:   Trichloroacetaldehyde hydrate
Image of Chloral hydrate
CAS Number:
Linear Formula:
 Cl3CCH(OH)2
Molecular Weight:
 165.40
 Beilstein/REAXYS Number:
 1698497
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Suitability
  Special Grade
  Quality clarifier
Show More