• USA Home
  • Search Results

  • chloralose309531587993311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloralose

1 Products
Synonym:   α-Chloralose, 1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose, Anhydro-D-glucochloral, Chloralose
Image of Chloralose
CAS Number:
Empirical Formula (Hill Notation):
 C8H11Cl3O6
Molecular Weight:
 309.53
 Beilstein/REAXYS Number:
 85418
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More