• USA Home
  • Search Results

  • chloraminethydrate22764anhydrousbasis14935873611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloramine-T hydrate

1 Products
Synonym:   N-Chloro-p-toluenesulfonamide sodium salt hydrate
Image of Chloramine-T hydrate
CAS Number:
Linear Formula:
 CH3C6H4SO2N(Cl)Na · xH2O
Molecular Weight:
 227.64 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
857319 Chloramine-T hydrate, 95%,