• USA Home
  • Search Results

  • chloraminettrihydrate28169708050411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloramine T trihydrate

7 Products
Synonym:   Chloramine T trihydrate, Tosylchloramide sodium, N-Chloro-4-toluenesulfonamide sodium salt, Chlorine bleaching solution
Image of Chloramine T trihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C7H7ClNNaO2S · 3H2O
Molecular Weight:
 281.69
 EC Index Number:
 204-854-7
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Quality clarifier
  Potency
  Bulk Density
  Solubility
  Special Grade
  pH Value
Show More