• USA Home
  • Search Results

  • chloramphenicol323135675711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloramphenicol

9 Products
Synonym:   D-(−)-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide, D-(−)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, D-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-4-nitrophenethyl]acetamide, Chloromycetin
Image of Chloramphenicol
CAS Number:
Linear Formula:
 Cl2CHCONHCH(CH2OH)CH(OH)C6H4NO2
Molecular Weight:
 323.13
 Beilstein/REAXYS Number:
 2225532
 EC Number:
 MDL number:
 NACRES:
 NA.76
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Agency/Method
  Anal. Standard Form
  Shelf Life
  pK-Values
  Product Line
  Manufacturer designation
  Assay
  Application(s)
  Mode of Action
  Special Grade
  Physical Form
  Solubility
Show More