• USA Home
  • Search Results

  • chlorhexidine505455556111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorhexidine

4 Products
Synonym:   1,1′-Hexamethylenebis[5-(4-chlorophenyl)biguanide]
Image of Chlorhexidine
CAS Number:
Linear Formula:
 [-(CH2)3NHC(=NH)NHC(=NH)NHC6H4Cl]2
Molecular Weight:
 505.45
 Beilstein/REAXYS Number:
 2826432
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Manufacturer designation
  Special Grade
Show More