• USA Home
  • Search Results

  • chlorhexidineacetate625555695111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorhexidine acetate

4 Products
Synonym:   Chlorhexidine diacetate salt, Chlorhexidine diacetate
CAS Number:
Empirical Formula (Hill Notation):
 C22H30Cl2N10 · 2C2H4O2
Molecular Weight:
 625.55
 EC Number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
Show More